missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-2-Aminomethyl-1-cyclohexanol, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 265890025

 View more versions of this product

Catalog No. AC265890025



Description

Description

Specifications

Specifications

trans-2-Aminomethyl-1-cyclohexanol
Authentic
1.4905 to 1.4925
Colorless
99+%
99% min. (GC)
OHC6H10CH2NH2
1r,2s---trans-2-aminomethyl cyclohexanol, 1r,2s-2-aminomethyl cyclohexan-1-ol, 1r,2s-2-aminomethyl cyclohexanol, 1r-2alpha-aminomethylcyclohexan-1beta-ol, 2-aminomethyl cyclohexanol, e, cyclohexanol, 2-aminomethyl-, trans, jgkfbzbvcawdfd-nkwvepmbsa-n, trans-2-aminomethyl cyclohexanol, trans-2-aminomethyl-1-cyclohexanol, trans-2-aminomethyl-cyclohexanol
JGKFBZBVCAWDFD-NKWVEPMBSA-O
2724658
Liquid After Melting
91°C
Glass bottle
226°C
2.5g
5691-09-8
C7H15NO
MFCD06213163
Solubility in water: soluble. Other solubilities: soluble in chloroform,ethanol,methanol
129.2
129.2
99+%

Chemical Identifiers

5691-09-8
129.2
JGKFBZBVCAWDFD-NKWVEPMBSA-O
2724658
C7H15NO
MFCD06213163
1r,2s---trans-2-aminomethyl cyclohexanol, 1r,2s-2-aminomethyl cyclohexan-1-ol, 1r,2s-2-aminomethyl cyclohexanol, 1r-2alpha-aminomethylcyclohexan-1beta-ol, 2-aminomethyl cyclohexanol, e, cyclohexanol, 2-aminomethyl-, trans, jgkfbzbvcawdfd-nkwvepmbsa-n, trans-2-aminomethyl cyclohexanol, trans-2-aminomethyl-1-cyclohexanol, trans-2-aminomethyl-cyclohexanol
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents