Please login to your online account to display your discounted pricing

trans-2-Aminomethyl-1-cyclohexanol, 99+%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  265890010

 View more versions of this product

Catalog No. AC265890010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Formula Weight 129.2
Physical Form Liquid After Melting
Quantity 1g
Color Clear colorless
CAS 1384887
Molecular Formula C7H15NO
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Refractive Index 1.4905 to 1.4925 (20°C, 589nm)
Chemical Name or Material trans-2-Aminomethyl-1-cyclohexanol
MDL Number MFCD06213163
Assay Percent Range 99+%
Name Note 99+%
Linear Formula OHC6H10CH2NH2
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 MOISTURE SENSITIVE

Specifications

Chemical Name or Material trans-2-Aminomethyl-1-cyclohexanol
Name Note 99+%
CAS 1384887
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C7H15NO
Linear Formula OHC6H10CH2NH2
Formula Weight 129.2
MDL Number MFCD06213163
Accessories

MOISTURE SENSITIVE

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents