Please login to your online account to display your discounted pricing

trans-2-Aminocyclohexanol hydrochloride, 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC206980250

 View more versions of this product

Catalog No. AC206980250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 151.64
Quantity 25g
Color White or beige to light brown
Melting Point 173° to 177°C
Beilstein 13, III, 704
Chemical Name or Material trans-2-Aminocyclohexanol hydrochloride
Packaging Glass bottle
Assay Percent Range 99%
Molecular Formula C6H13NO·HCl
MDL Number MFCD00001490
CAS 5456-63-3
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula H2NC6H10OH·HCl
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

Specifications

Chemical Name or Material trans-2-Aminocyclohexanol hydrochloride
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C6H13NO·HCl
Linear Formula H2NC6H10OH·HCl
Formula Weight 151.64
MDL Number MFCD00001490
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents