missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-2-Amino-1-cyclohexanecarboxylic acid, 98%, ACROS Organics™

Chemical Identifiers

CAS 5691-19-0
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.19
MDL Number MFCD00145418
InChI Key USQHEVWOPJDAAX-PHDIDXHHSA-N
Synonym 1r,2r-2-aminocyclohexane-1-carboxylic acid, 1r,2r-2-aminocyclohexanecarboxylic acid, 2-aminocyclohexanecarboxylic acid #, 2-nchca, cyclohexanecarboxylic acid, 2-amino-, 1r,2r, cyclohexanecarboxylic acid, 2-amino-, 1r,2r-rel, cyclohexanecarboxylic acid, 2-amino-, trans, trans-2-amino-1-cyclohexanecarboxylic acid, trans-2-amino-cyclohexanecarboxylic acid, trans-2-aminocyclohexanecarboxylic acid
PubChem CID 2724640
IUPAC Name (1R,2R)-2-aminocyclohexane-1-carboxylic acid
SMILES C1CCC(C(C1)C(=O)O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC265630010
SDS View Documents
Thermo Fisher Scientific
265630010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC265630050
SDS View Documents
Thermo Fisher Scientific
AC265630050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

5691-19-0
143.19
USQHEVWOPJDAAX-PHDIDXHHSA-N
2724640
C1CCC(C(C1)C(=O)O)N
C7H13NO2
MFCD00145418
1r,2r-2-aminocyclohexane-1-carboxylic acid, 1r,2r-2-aminocyclohexanecarboxylic acid, 2-aminocyclohexanecarboxylic acid #, 2-nchca, cyclohexanecarboxylic acid, 2-amino-, 1r,2r, cyclohexanecarboxylic acid, 2-amino-, 1r,2r-rel, cyclohexanecarboxylic acid, 2-amino-, trans, trans-2-amino-1-cyclohexanecarboxylic acid, trans-2-amino-cyclohexanecarboxylic acid, trans-2-aminocyclohexanecarboxylic acid
(1R,2R)-2-aminocyclohexane-1-carboxylic acid

Specifications

Authentic
White
1g
98%
MFCD00145418
Solubility in water: soluble in water. Other solubilities: soluble in methanol
C1CCC(C(C1)C(=O)O)N
143.19
143.19
98%
Glass bottle
274°C to 278°C
5691-19-0
C7H13NO2
1r,2r-2-aminocyclohexane-1-carboxylic acid, 1r,2r-2-aminocyclohexanecarboxylic acid, 2-aminocyclohexanecarboxylic acid #, 2-nchca, cyclohexanecarboxylic acid, 2-amino-, 1r,2r, cyclohexanecarboxylic acid, 2-amino-, 1r,2r-rel, cyclohexanecarboxylic acid, 2-amino-, trans, trans-2-amino-1-cyclohexanecarboxylic acid, trans-2-amino-cyclohexanecarboxylic acid, trans-2-aminocyclohexanecarboxylic acid
USQHEVWOPJDAAX-PHDIDXHHSA-N
(1R,2R)-2-aminocyclohexane-1-carboxylic acid
2724640
Powder and Chunks
trans-2-Amino-1-cyclohexanecarboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS