Learn More
trans-2-Amino-1-cyclohexanecarboxylic acid, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 265630010
Specifications
trans-2-Amino-1-cyclohexanecarboxylic acid | |
Glass bottle | |
274°C to 278°C | |
5691-19-0 | |
C7H13NO2 | |
1r,2r-2-aminocyclohexane-1-carboxylic acid, 1r,2r-2-aminocyclohexanecarboxylic acid, 2-aminocyclohexanecarboxylic acid #, 2-nchca, cyclohexanecarboxylic acid, 2-amino-, 1r,2r, cyclohexanecarboxylic acid, 2-amino-, 1r,2r-rel, cyclohexanecarboxylic acid, 2-amino-, trans, trans-2-amino-1-cyclohexanecarboxylic acid, trans-2-amino-cyclohexanecarboxylic acid, trans-2-aminocyclohexanecarboxylic acid | |
USQHEVWOPJDAAX-PHDIDXHHSA-N | |
(1R,2R)-2-aminocyclohexane-1-carboxylic acid | |
2724640 | |
Powder and Chunks |
Authentic | |
White | |
1g | |
98% | |
MFCD00145418 | |
Solubility in water: soluble in water. Other solubilities: soluble in methanol | |
C1CCC(C(C1)C(=O)O)N | |
143.19 | |
143.19 | |
98% |
Chemical Identifiers
5691-19-0 | |
143.19 | |
USQHEVWOPJDAAX-PHDIDXHHSA-N | |
2724640 | |
C1CCC(C(C1)C(=O)O)N |
C7H13NO2 | |
MFCD00145418 | |
1r,2r-2-aminocyclohexane-1-carboxylic acid, 1r,2r-2-aminocyclohexanecarboxylic acid, 2-aminocyclohexanecarboxylic acid #, 2-nchca, cyclohexanecarboxylic acid, 2-amino-, 1r,2r, cyclohexanecarboxylic acid, 2-amino-, 1r,2r-rel, cyclohexanecarboxylic acid, 2-amino-, trans, trans-2-amino-1-cyclohexanecarboxylic acid, trans-2-amino-cyclohexanecarboxylic acid, trans-2-aminocyclohexanecarboxylic acid | |
(1R,2R)-2-aminocyclohexane-1-carboxylic acid |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning