Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-2,5-Dimethylpiperazine, 98%

Chemical Identifiers

Chemical Name or Material trans-2,5-Dimethylpiperazine
CAS 2815-34-1
Molecular Formula C6H14N2
Formula Weight 114.19
MDL Number MFCD00070516
UN Number UN2925
Beilstein 79886
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2315006

  SDS
alfa aesar™
B2315006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2315014

  SDS
alfa aesar™
B2315014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2315022

  SDS
alfa aesar™
B2315022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 114.19
Flash Point 58°C (136°F)
RTECS Number TL5950000
MDL Number MFCD00070516
Chemical Name or Material trans-2,5-Dimethylpiperazine
Melting Point 115° to 119°C
UN Number UN2925
Assay 98%
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.
Beilstein 79886
Molecular Formula C6H14N2
Quantity 5g
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 2815-34-1
Boiling Point 162° to 165°C
Health Hazard 2 GHS H Statement
H314-H318-H228-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
Harmful in contact with skin.
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H228-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
Harmful in contact with skin.