Learn More
trans-2,5-Dimethylpiperazine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2315022
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
trans-2,5-Dimethylpiperazine | |
115°C to 119°C | |
58°C (136°F) | |
MFCD00070516 | |
UN2925 | |
trans-2,5-dimethylpiperazine, 2r,5s-2,5-dimethylpiperazine, 2s,5r-2,5-dimethylpiperazine, 2s,5r-2,5-dimethyl-piperazine, piperazine, 2,5-dimethyl-, 2r,5s-rel, 2r,5s-2,5-dimethylpiperazin, piperazine, 2,5-dimethyl-, trans, 2r,5s-2,5-dimethyl-piperazine, pubchem15878, 2,5-dimethylpiperazine # | |
CC1CNC(CN1)C | |
114.192 | |
114.19 |
2815-34-1 | |
162°C to 165°C | |
C6H14N2 | |
100 g | |
79886 | |
NSMWYRLQHIXVAP-OLQVQODUSA-N | |
(2R,5S)-2,5-dimethylpiperazine | |
220672 | |
98% |
Chemical Identifiers
2815-34-1 | |
114.192 | |
NSMWYRLQHIXVAP-OLQVQODUSA-N | |
220672 | |
CC1CNC(CN1)C |
C6H14N2 | |
MFCD00070516 | |
trans-2,5-dimethylpiperazine, 2r,5s-2,5-dimethylpiperazine, 2s,5r-2,5-dimethylpiperazine, 2s,5r-2,5-dimethyl-piperazine, piperazine, 2,5-dimethyl-, 2r,5s-rel, 2r,5s-2,5-dimethylpiperazin, piperazine, 2,5-dimethyl-, trans, 2r,5s-2,5-dimethyl-piperazine, pubchem15878, 2,5-dimethylpiperazine # | |
(2R,5S)-2,5-dimethylpiperazine |
Safety and Handling
GHS H Statement
H314-H318-H228-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
Harmful in contact with skin.
P210-P240-P241-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H228-H312-H314-H500
DOTInformation : Hazard Class: 4.1; Packaging Group: II
EINECSNumber : 000-000-0
RTECSNumber : TL5950000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only