Please login to your online account to display your discounted pricing

Alfa Aesar™ (±)-trans-2,5-Bis-(3,4,5-trimethoxyphenyl)-1,3-dioxolane

Chemical Identifiers

Chemical Name or Material (±)-trans-2,5-Bis-(3,4,5-trimethoxyphenyl)-1,3-dioxolane
CAS 116673-45-1
Formula Weight 406.43
MDL Number MFCD00155367
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67356LB0

 
alfa aesar™
J67356LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67356MCR

 
alfa aesar™
J67356MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 406.43
MDL Number MFCD00155367
Quantity 5mg
Color White
Chemical Name or Material (±)-trans-2,5-Bis-(3,4,5-trimethoxyphenyl)-1,3-dioxolane
Physical Form Solid
CAS 116673-45-1