missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-2,4-Difluorocinnamic acid, 97%

$129.61 - $129.61

Chemical Identifiers

CAS 94977-52-3
Molecular Formula C9H6F2O2
Formula Weight 184.14
MDL Number MFCD00010317
Beilstein 3129944
Chemical Name or Material trans-2,4-Difluorocinnamic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1827706 SDS Alfa Aesar™
A1827706
5g Each for $129.61
AAA1827714 SDS Alfa Aesar™
A1827714
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

94977-52-3
184.14
3129944
C9H6F2O2
MFCD00010317
trans-2,4-Difluorocinnamic acid

Specifications

94977-52-3
184.14
3129944
5g
97%
C9H6F2O2
MFCD00010317
215° to 219°C
trans-2,4-Difluorocinnamic acid

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning