Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-2,4-Dichlorocinnamic acid, 98%

$36.61 - $367.65

Chemical Identifiers

Chemical Name or Material trans-2,4-Dichlorocinnamic acid
CAS 20595-45-3
Molecular Formula C9H6Cl2O2
Formula Weight 217.05
MDL Number MFCD00004373
Beilstein 1369152
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1382406

  SDS
alfa aesar™
A1382406
5g Each for $36.61

AAA1382414

  SDS
alfa aesar™
A1382414
25g Each for $113.01

AAA1382422

  SDS
alfa aesar™
A1382422
100g Each for $367.65
Description & Specifications

Specifications

Formula Weight 217.05
Chemical Name or Material trans-2,4-Dichlorocinnamic acid
Assay 98%
Molecular Formula C9H6Cl2O2
MDL Number MFCD00004373
CAS 20595-45-3
Beilstein 1369152
Melting Point 233° to 235°C
Health Hazard 1 Warning
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number GD8575000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.