Learn More
trans-2-(4-chlorophenyl)vinylboronic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 431340100
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
trans-2-(4-chlorophenyl)vinylboronic acid | |
Authentic | |
Glass bottle | |
MFCD02093767 | |
trans-2-4-chlorophenyl vinylboronic acid, 4-chlorostyryl boronic acid, e-2-4-chlorophenyl ethenyl boronic acid, e-4-chlorostyrylboronic acid, boronic acid, 1e-2-4-chlorophenyl ethenyl, e-2-4-chlorophenyl ethenylboronic acid, trans-2-4-chlorophenyl ethenylboronic acid, rarechem al ba 0169, e-2-4-chlorophenyl vinyl boronic acid, 4-chlorostyrylboronic acid | |
OB(O)\C=C\C1=CC=C(Cl)C=C1 | |
182.41 | |
182.41 |
154230-29-2 | |
96% min. (HPLC) | |
C8H8BClO2 | |
10g | |
HWSDRAPTZRYXHN-AATRIKPKSA-N | |
[(E)-2-(4-chlorophenyl)ethenyl]boronic acid | |
642694 | |
97% |
Chemical Identifiers
154230-29-2 | |
182.41 | |
HWSDRAPTZRYXHN-AATRIKPKSA-N | |
642694 | |
OB(O)\C=C\C1=CC=C(Cl)C=C1 |
C8H8BClO2 | |
MFCD02093767 | |
trans-2-4-chlorophenyl vinylboronic acid, 4-chlorostyryl boronic acid, e-2-4-chlorophenyl ethenyl boronic acid, e-4-chlorostyrylboronic acid, boronic acid, 1e-2-4-chlorophenyl ethenyl, e-2-4-chlorophenyl ethenylboronic acid, trans-2-4-chlorophenyl ethenylboronic acid, rarechem al ba 0169, e-2-4-chlorophenyl vinyl boronic acid, 4-chlorostyrylboronic acid | |
[(E)-2-(4-chlorophenyl)ethenyl]boronic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
RUO â Research Use Only