Please login to your online account to display your discounted pricing

trans-2-(4-chlorophenyl)vinylboronic acid, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC431340010

 View more versions of this product

Catalog No. AC431340010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥96.0%
Formula Weight 182.41
Quantity 1g
Color White to off-white
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 97%
Chemical Name or Material trans-2-(4-chlorophenyl)vinylboronic acid
CAS 154230-29-2
MDL Number MFCD02093767
Molecular Formula C8H8BClO2
Infrared Spectrum Authentic

Specifications

Chemical Name or Material trans-2-(4-chlorophenyl)vinylboronic acid
CAS 154230-29-2
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H8BClO2
Formula Weight 182.41
MDL Number MFCD02093767
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents