Please login to your online account to display your discounted pricing

trans-2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)styrene, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC432030050

 View more versions of this product

Catalog No. AC432030050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥94.0%
Formula Weight 230.11
Quantity 5g
Color Clear colorless to pale yellow
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material trans-2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)styrene
Assay Percent Range 97%
Molecular Formula C14H19BO2
MDL Number MFCD03453666
CAS 83947-56-2
Synonym trans-2-Phenylvinylboronic acid pinacol ester
Infrared Spectrum Authentic
Packaging Glass bottle

Specifications

Chemical Name or Material trans-2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)styrene
CAS 83947-56-2
Assay ≥94.0%
Assay Percent Range 97%
Molecular Formula C14H19BO2
Formula Weight 230.11
MDL Number MFCD03453666
Synonym trans-2-Phenylvinylboronic acid pinacol ester
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents