Learn More
trans-2,3-Dimethylacrylic Acid, 98%, Thermo Scientific Chemicals
$172.31 - $172.31
Chemical Identifiers
CAS | 80-59-1 |
---|---|
Molecular Formula | C5H8O2 |
Molecular Weight (g/mol) | 100.12 |
MDL Number | MFCD00066864 |
InChI Key | UIERETOOQGIECD-ONEGZZNKSA-N |
Synonym | tiglic acid, tiglinic acid, cevadic acid, 2-methyl-2-butenoic acid, trans-2-methylcrotonic acid, trans-2,3-dimethylacrylic acid, 2-methylbut-2-enoic acid, e-2-methyl-2-butenoic acid, trans-2-methyl-2-butenoic acid, e-2,3-dimethylacrylic acid |
PubChem CID | 125468 |
ChEBI | CHEBI:9592 |
IUPAC Name | (E)-2-methylbut-2-enoic acid |
SMILES | C\C=C(/C)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC138971000
|
Thermo Scientific Chemicals
138971000 |
100 g | Glass bottle |
Each for $172.31
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
80-59-1 | |
100.12 | |
UIERETOOQGIECD-ONEGZZNKSA-N | |
125468 | |
(E)-2-methylbut-2-enoic acid |
C5H8O2 | |
MFCD00066864 | |
tiglic acid, tiglinic acid, cevadic acid, 2-methyl-2-butenoic acid, trans-2-methylcrotonic acid, trans-2,3-dimethylacrylic acid, 2-methylbut-2-enoic acid, e-2-methyl-2-butenoic acid, trans-2-methyl-2-butenoic acid, e-2,3-dimethylacrylic acid | |
CHEBI:9592 | |
C\C=C(/C)C(O)=O |
Specifications
80-59-1 | |
White to Beige | |
198.4°C | |
Authentic | |
Glass bottle | |
CH3CH=C(CH3)CO2H | |
100 g | |
0.96 | |
tiglic acid, tiglinic acid, cevadic acid, 2-methyl-2-butenoic acid, trans-2-methylcrotonic acid, trans-2,3-dimethylacrylic acid, 2-methylbut-2-enoic acid, e-2-methyl-2-butenoic acid, trans-2-methyl-2-butenoic acid, e-2,3-dimethylacrylic acid | |
UIERETOOQGIECD-ONEGZZNKSA-N | |
(E)-2-methylbut-2-enoic acid | |
125468 | |
100.12 | |
Crystalline Powder and/or Chunks |
61.0°C to 65.0°C | |
0.9600g/mL | |
95°C | |
98% | |
C5H8O2 | |
MFCD00066864 | |
02, 430 | |
15, 9589 | |
Solubility in water: soluble in hot water. Other solubilities: 1g/10mL methanol,soluble in alcohol and ether | |
C\C=C(/C)C(O)=O | |
100.12 | |
CHEBI:9592 | |
98% | |
trans-2, 3-Dimethylacrylic acid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Danger
EINECSNumber : 201-295-
RUO – Research Use Only