Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-2,3-Dibromo-2-butene-1,4-diol, 97%

$115.68 - $455.28

Chemical Identifiers

Chemical Name or Material trans-2,3-Dibromo-2-butene-1,4-diol
CAS 21285-46-1
Molecular Formula C4H6Br2O2
Formula Weight 245.91g/mol
MDL Number MFCD00004698
Beilstein 1720775
Synonym trans-1,4-Dihydroxy-2,3-dibromo-2-butene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2163722

 
alfa aesar™
B21637-22
100g Each for $115.68

AAB2163736

 
alfa aesar™
B21637-36
500g Each for $455.28
Description & Specifications

Specifications

Melting Point 111° to 115°C
Quantity 100g
Chemical Name or Material trans-2,3-Dibromo-2-butene-1,4-diol
CAS 21285-46-1
Assay 97%
Molecular Formula C4H6Br2O2
Formula Weight 245.91g/mol
MDL Number MFCD00004698
Beilstein 1720775
Synonym trans-1,4-Dihydroxy-2,3-dibromo-2-butene
RTECS Number EM6910000
TSCA TSCA