missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-2,3,4-Trimethoxycinnamic acid, 99%

$57.36 - $95.92

Chemical Identifiers

CAS 33130-03-9
Molecular Formula C12H14O5
Molecular Weight (g/mol) 238.239
MDL Number MFCD00014376
InChI Key ZYOPDNLIHHFGEC-FNORWQNLSA-N
Synonym 2,3,4-trimethoxycinnamic acid, trans-2,3,4-trimethoxycinnamic acid, 3-2,3,4-trimethoxyphenyl acrylic acid, 2e-3-2,3,4-trimethoxyphenyl prop-2-enoic acid, 2-propenoic acid, 3-2,3,4-trimethoxyphenyl, 3-2,3,4-trimethoxyphenyl propenoic acid, trans-2, 3, 4-trimethoxycinnamate, 2e-3-2,3,4-trimethoxyphenyl-2-propenoic acid, e-3-2,3,4-trimethoxyphenyl acrylic acid, 2e-3-2,3,4-trimethoxyphenyl acrylic acid
PubChem CID 735841
IUPAC Name (E)-3-(2,3,4-trimethoxyphenyl)prop-2-enoic acid
SMILES COC1=C(C(=C(C=C1)C=CC(=O)O)OC)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0170306 View Documents Alfa Aesar™
L0170306
5g Each for $57.36
 
AAL0170314 View Documents Alfa Aesar™
L0170314
25g Each for $95.92
 
Specifications

Chemical Identifiers

33130-03-9
238.239
ZYOPDNLIHHFGEC-FNORWQNLSA-N
735841
COC1=C(C(=C(C=C1)C=CC(=O)O)OC)OC
C12H14O5
MFCD00014376
2,3,4-trimethoxycinnamic acid, trans-2,3,4-trimethoxycinnamic acid, 3-2,3,4-trimethoxyphenyl acrylic acid, 2e-3-2,3,4-trimethoxyphenyl prop-2-enoic acid, 2-propenoic acid, 3-2,3,4-trimethoxyphenyl, 3-2,3,4-trimethoxyphenyl propenoic acid, trans-2, 3, 4-trimethoxycinnamate, 2e-3-2,3,4-trimethoxyphenyl-2-propenoic acid, e-3-2,3,4-trimethoxyphenyl acrylic acid, 2e-3-2,3,4-trimethoxyphenyl acrylic acid
(E)-3-(2,3,4-trimethoxyphenyl)prop-2-enoic acid

Specifications

172°C to 176°C
33130-03-9
MFCD00014376
2,3,4-trimethoxycinnamic acid, trans-2,3,4-trimethoxycinnamic acid, 3-2,3,4-trimethoxyphenyl acrylic acid, 2e-3-2,3,4-trimethoxyphenyl prop-2-enoic acid, 2-propenoic acid, 3-2,3,4-trimethoxyphenyl, 3-2,3,4-trimethoxyphenyl propenoic acid, trans-2, 3, 4-trimethoxycinnamate, 2e-3-2,3,4-trimethoxyphenyl-2-propenoic acid, e-3-2,3,4-trimethoxyphenyl acrylic acid, 2e-3-2,3,4-trimethoxyphenyl acrylic acid
COC1=C(C(=C(C=C1)C=CC(=O)O)OC)OC
238.239
238.24
trans-2,3,4-Trimethoxycinnamic acid
5g
C12H14O5
2657904
ZYOPDNLIHHFGEC-FNORWQNLSA-N
(E)-3-(2,3,4-trimethoxyphenyl)prop-2-enoic acid
735841
99%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning