missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
trans-2,3,4-Trimethoxycinnamic acid, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals L0170306
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
trans-2,3,4-Trimethoxycinnamic acid | |
172°C to 176°C | |
MFCD00014376 | |
2657904 | |
ZYOPDNLIHHFGEC-FNORWQNLSA-N | |
(E)-3-(2,3,4-trimethoxyphenyl)prop-2-enoic acid | |
735841 | |
99% |
33130-03-9 | |
C12H14O5 | |
5 g | |
2,3,4-trimethoxycinnamic acid, trans-2,3,4-trimethoxycinnamic acid, 3-2,3,4-trimethoxyphenyl acrylic acid, 2e-3-2,3,4-trimethoxyphenyl prop-2-enoic acid, 2-propenoic acid, 3-2,3,4-trimethoxyphenyl, 3-2,3,4-trimethoxyphenyl propenoic acid, trans-2, 3, 4-trimethoxycinnamate, 2e-3-2,3,4-trimethoxyphenyl-2-propenoic acid, e-3-2,3,4-trimethoxyphenyl acrylic acid, 2e-3-2,3,4-trimethoxyphenyl acrylic acid | |
COC1=C(C(=C(C=C1)C=CC(=O)O)OC)OC | |
238.239 | |
238.24 |
Chemical Identifiers
33130-03-9 | |
238.239 | |
ZYOPDNLIHHFGEC-FNORWQNLSA-N | |
735841 | |
COC1=C(C(=C(C=C1)C=CC(=O)O)OC)OC |
C12H14O5 | |
MFCD00014376 | |
2,3,4-trimethoxycinnamic acid, trans-2,3,4-trimethoxycinnamic acid, 3-2,3,4-trimethoxyphenyl acrylic acid, 2e-3-2,3,4-trimethoxyphenyl prop-2-enoic acid, 2-propenoic acid, 3-2,3,4-trimethoxyphenyl, 3-2,3,4-trimethoxyphenyl propenoic acid, trans-2, 3, 4-trimethoxycinnamate, 2e-3-2,3,4-trimethoxyphenyl-2-propenoic acid, e-3-2,3,4-trimethoxyphenyl acrylic acid, 2e-3-2,3,4-trimethoxyphenyl acrylic acid | |
(E)-3-(2,3,4-trimethoxyphenyl)prop-2-enoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 251-388-5
TSCA : No
Recommended Storage : Ambient temperatures