missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-(1R,2R)-(-)-N,N'-Dimethyl-1,2-cyclohexanediamine dihydrochloride, 98%, ACROS Organics™

Chemical Identifiers

CAS 70708-33-7
Molecular Formula C8H18N2·2ClH
Molecular Weight (g/mol) 215.17
InChI Key UDYXZFBJGBDHHP-RHJRFJOKSA-N
Synonym 1,2-cyclohexanediamine, n,n'-dimethyl-, dihydrochloride, 1r-trans, 1r,2r-1-n,2-n-dimethylcyclohexane-1,2-diamine dihydrochloride, 1r,2r-n,n'-dimethyl-1,2-cyclohexanediamine dihydrochloride, 1r,2r-n1,n2-dimethylcyclohexane-1,2-diamine dihydrochloride, trans-1r,2r---n,n'-dimethyl-1,2-cyclohexanediamine dihydrochloride, trans-1r,2r-n,n'-bismethyl-1,2-cyclohexane diamine dihydrochloride, trans-1r,2r-n,n'-bismethyl-1,2-cyclohexanediamine 2hcl, trans-1r,2r-n,n'-bismethyl-1,2-cyclohexanediamine dihydrochloride, trans-n,n'-dimethyl-1,2-diaminocyclohexane dihydrochloride, trans-n1,n2-dimethylcyclohexane-1,2-diamine dihydrochloride
PubChem CID 44890801
IUPAC Name (1R,2R)-1-N,2-N-dimethylcyclohexane-1,2-diamine;dihydrochloride
SMILES CNC1CCCCC1NC.Cl.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC451020010
SDS View Documents
Thermo Fisher Scientific
AC451020010
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC451020050
SDS View Documents
Thermo Fisher Scientific
AC451020050
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC451020250
SDS View Documents
Thermo Fisher Scientific
AC451020250
25g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

70708-33-7
215.17
1,2-cyclohexanediamine, n,n'-dimethyl-, dihydrochloride, 1r-trans, 1r,2r-1-n,2-n-dimethylcyclohexane-1,2-diamine dihydrochloride, 1r,2r-n,n'-dimethyl-1,2-cyclohexanediamine dihydrochloride, 1r,2r-n1,n2-dimethylcyclohexane-1,2-diamine dihydrochloride, trans-1r,2r---n,n'-dimethyl-1,2-cyclohexanediamine dihydrochloride, trans-1r,2r-n,n'-bismethyl-1,2-cyclohexane diamine dihydrochloride, trans-1r,2r-n,n'-bismethyl-1,2-cyclohexanediamine 2hcl, trans-1r,2r-n,n'-bismethyl-1,2-cyclohexanediamine dihydrochloride, trans-n,n'-dimethyl-1,2-diaminocyclohexane dihydrochloride, trans-n1,n2-dimethylcyclohexane-1,2-diamine dihydrochloride
(1R,2R)-1-N,2-N-dimethylcyclohexane-1,2-diamine;dihydrochloride
C8H18N2·2ClH
UDYXZFBJGBDHHP-RHJRFJOKSA-N
44890801
CNC1CCCCC1NC.Cl.Cl

Specifications

Glass Bottle
70708-33-7
C8H18N2·2ClH
UDYXZFBJGBDHHP-RHJRFJOKSA-N
(1R,2R)-1-N,2-N-dimethylcyclohexane-1,2-diamine;dihydrochloride
44890801
98%
1g
97.5 to max. 102.5 % min. (Argentometry)
1,2-cyclohexanediamine, n,n'-dimethyl-, dihydrochloride, 1r-trans, 1r,2r-1-n,2-n-dimethylcyclohexane-1,2-diamine dihydrochloride, 1r,2r-n,n'-dimethyl-1,2-cyclohexanediamine dihydrochloride, 1r,2r-n1,n2-dimethylcyclohexane-1,2-diamine dihydrochloride, trans-1r,2r---n,n'-dimethyl-1,2-cyclohexanediamine dihydrochloride, trans-1r,2r-n,n'-bismethyl-1,2-cyclohexane diamine dihydrochloride, trans-1r,2r-n,n'-bismethyl-1,2-cyclohexanediamine 2hcl, trans-1r,2r-n,n'-bismethyl-1,2-cyclohexanediamine dihydrochloride, trans-n,n'-dimethyl-1,2-diaminocyclohexane dihydrochloride, trans-n1,n2-dimethylcyclohexane-1,2-diamine dihydrochloride
CNC1CCCCC1NC.Cl.Cl
215.17
215.17
trans-(1R,2R)-(-)-N,N'-Dimethyl-1,2-cyclohexanediamine dihydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist

GHS Signal Word: Warning