missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
trans-(1R,2R)-(-)-N,N'-Dimethyl-1,2-cyclohexanediamine dihydrochloride, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC451020250
Specifications
trans-(1R,2R)-(-)-N,N'-Dimethyl-1,2-cyclohexanediamine dihydrochloride | |
70708-33-7 | |
C8H18N2·2ClH | |
UDYXZFBJGBDHHP-RHJRFJOKSA-N | |
(1R,2R)-1-N,2-N-dimethylcyclohexane-1,2-diamine;dihydrochloride | |
44890801 | |
98% |
25g | |
97.5 to max. 102.5% min. (Argentometry) | |
1,2-cyclohexanediamine, n,n'-dimethyl-, dihydrochloride, 1r-trans, 1r,2r-1-n,2-n-dimethylcyclohexane-1,2-diamine dihydrochloride, 1r,2r-n,n'-dimethyl-1,2-cyclohexanediamine dihydrochloride, 1r,2r-n1,n2-dimethylcyclohexane-1,2-diamine dihydrochloride, trans-1r,2r---n,n'-dimethyl-1,2-cyclohexanediamine dihydrochloride, trans-1r,2r-n,n'-bismethyl-1,2-cyclohexane diamine dihydrochloride, trans-1r,2r-n,n'-bismethyl-1,2-cyclohexanediamine 2hcl, trans-1r,2r-n,n'-bismethyl-1,2-cyclohexanediamine dihydrochloride, trans-n,n'-dimethyl-1,2-diaminocyclohexane dihydrochloride, trans-n1,n2-dimethylcyclohexane-1,2-diamine dihydrochloride | |
CNC1CCCCC1NC.Cl.Cl | |
215.17 | |
215.17 |
Chemical Identifiers
70708-33-7 | |
215.17 | |
1,2-cyclohexanediamine, n,n'-dimethyl-, dihydrochloride, 1r-trans, 1r,2r-1-n,2-n-dimethylcyclohexane-1,2-diamine dihydrochloride, 1r,2r-n,n'-dimethyl-1,2-cyclohexanediamine dihydrochloride, 1r,2r-n1,n2-dimethylcyclohexane-1,2-diamine dihydrochloride, trans-1r,2r---n,n'-dimethyl-1,2-cyclohexanediamine dihydrochloride, trans-1r,2r-n,n'-bismethyl-1,2-cyclohexane diamine dihydrochloride, trans-1r,2r-n,n'-bismethyl-1,2-cyclohexanediamine 2hcl, trans-1r,2r-n,n'-bismethyl-1,2-cyclohexanediamine dihydrochloride, trans-n,n'-dimethyl-1,2-diaminocyclohexane dihydrochloride, trans-n1,n2-dimethylcyclohexane-1,2-diamine dihydrochloride | |
(1R,2R)-1-N,2-N-dimethylcyclohexane-1,2-diamine;dihydrochloride |
C8H18N2·2ClH | |
UDYXZFBJGBDHHP-RHJRFJOKSA-N | |
44890801 | |
CNC1CCCCC1NC.Cl.Cl |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist
GHS Signal Word: Warning