missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-1-Methyl-4-carboxy-5-(3-pyridyl)-2-pyrrolidinone, TCI America™

$25.97 - $235.28

Chemical Identifiers

CAS 33224-01-0
Molecular Formula C11H12N2O3
Molecular Weight (g/mol) 220.228
MDL Number MFCD00075394
InChI Key DEYLVDCFTICBTB-WCBMZHEXSA-N
Synonym trans-4′-Carboxycotinine
PubChem CID 671220
IUPAC Name (2S,3S)-1-methyl-5-oxo-2-pyridin-3-ylpyrrolidine-3-carboxylic acid
SMILES CN1C(C(CC1=O)C(=O)O)C2=CN=CC=C2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M1053100MG View Documents TCI America
M1053100MG
100mg Each for $25.97
 
M10531G View Documents TCI America
M10531G
1g Each for $79.45
 
M10535G View Documents TCI America
M10535G
5g Each for $235.28
 
Specifications

Chemical Identifiers

33224-01-0
220.228
DEYLVDCFTICBTB-WCBMZHEXSA-N
671220
CN1C(C(CC1=O)C(=O)O)C2=CN=CC=C2
C11H12N2O3
MFCD00075394
trans-4′-Carboxycotinine
(2S,3S)-1-methyl-5-oxo-2-pyridin-3-ylpyrrolidine-3-carboxylic acid

Specifications

White-Yellow
100mg
C11H12N2O3
trans-4′C-Carboxycotinine
CN1C(C(CC1=O)C(=O)O)C2=CN=CC=C2
220.228
220.23
≥95.0% (GC,T)
194°C
33224-01-0
MFCD00075394
DEYLVDCFTICBTB-WCBMZHEXSA-N
(2S,3S)-1-methyl-5-oxo-2-pyridin-3-ylpyrrolidine-3-carboxylic acid
671220
Crystal-Powder at 20°C
trans-1-Methyl-4-carboxy-5-(3-pyridyl)-2-pyrrolidinone
Safety and Handling

Safety and Handling

TSCA : No