Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-1-Hepten-1-ylboronic acid pinacol ester, 95%

$198.57 - $198.57

Chemical Identifiers

Chemical Name or Material trans-1-Hepten-1-ylboronic acid pinacol ester
CAS 169339-75-7
Molecular Formula C13H25BO2
Formula Weight 224.15
MDL Number MFCD05663884
Synonym E-2-(1-Heptenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2787606

 
alfa aesar™
H2787606
5g Each for $198.57
Description & Specifications

Specifications

Formula Weight 224.15
Chemical Name or Material trans-1-Hepten-1-ylboronic acid pinacol ester
Assay 95%
Molecular Formula C13H25BO2
MDL Number MFCD05663884
CAS 169339-75-7
Synonym E-2-(1-Heptenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Boiling Point 73° to 77°C (0.5mmHg)
Quantity 5g
Flash Point 106°C (223°F)