missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-1-Fluoro-4-(4-n-pentylcyclohexyl)benzene, 97%

Manufacturer:  Alfa Aesar™ H5525506

 View more versions of this product

Catalog No. AAH5525506



Specifications

Specifications

trans-1-Fluoro-4-(4-n-pentylcyclohexyl)benzene
5g
C17H25F
1-fluoro-4-1s,4r-4-pentylcyclohexyl benzene
CCCCCC1CCC(CC1)C2=CC=C(C=C2)F
248.385
248.38
34°C to 36°C
76802-61-4
MFCD11053471
VCIVJVKSOMDCMN-UHFFFAOYSA-N
1-fluoro-4-(4-pentylcyclohexyl)benzene
578937
97%

Chemical Identifiers

76802-61-4
248.385
VCIVJVKSOMDCMN-UHFFFAOYSA-N
578937
CCCCCC1CCC(CC1)C2=CC=C(C=C2)F
C17H25F
MFCD11053471
1-fluoro-4-1s,4r-4-pentylcyclohexyl benzene
1-fluoro-4-(4-pentylcyclohexyl)benzene
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents