missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-1-Fluoro-4-(4-n-heptylcyclohexyl)benzene, 97%

Manufacturer:  Alfa Aesar™ H5527403

 View more versions of this product

Catalog No. AAH5527403



Description

Description

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

trans-1-Fluoro-4-(4-n-heptylcyclohexyl)benzene
1g
C19H29F
1-fluoro-4-4-heptylcyclohexyl benzene
CCCCCCCC1CCC(CC1)C2=CC=C(C=C2)F
276.439
276.44
35°C to 37°C
76802-59-0
MFCD11053424
ZPNZXIUICNXXNC-UHFFFAOYSA-N
1-fluoro-4-(4-heptylcyclohexyl)benzene
578935
97%

Chemical Identifiers

76802-59-0
276.439
ZPNZXIUICNXXNC-UHFFFAOYSA-N
578935
CCCCCCCC1CCC(CC1)C2=CC=C(C=C2)F
C19H29F
MFCD11053424
1-fluoro-4-4-heptylcyclohexyl benzene
1-fluoro-4-(4-heptylcyclohexyl)benzene
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents