Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-1-Cinnamylpiperazine, 97%

$54.03 - $183.18

Chemical Identifiers

Chemical Name or Material trans-1-Cinnamylpiperazine
CAS 87179-40-6
Molecular Formula C13H18N2
Formula Weight 202.3
MDL Number MFCD00063251
Beilstein 608984
Synonym (E)-1-Phenyl-3-(1-piperazinyl)-1-propene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2476609

 
alfa aesar™
B2476609
10g Each for $54.03

AAB2476618

 
alfa aesar™
B2476618
50g Each for $183.18
Description & Specifications

Specifications

Formula Weight 202.3
Chemical Name or Material trans-1-Cinnamylpiperazine
Assay 97%
Molecular Formula C13H18N2
MDL Number MFCD00063251
CAS 87179-40-6
Beilstein 608984
Synonym (E)-1-Phenyl-3-(1-piperazinyl)-1-propene
Melting Point 37° to 40°C
Boiling Point 129°C (1mmHg)
Health Hazard 1 Warning
Quantity 10g
Flash Point >110°C (230°F)
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.