missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-1-(Boc-amino)-4-vinylcyclohexane, 97%, Thermo Scientific Chemicals

$305.10 - $1102.81

Chemical Identifiers

CAS 1198355-16-6
Molecular Formula C13H23NO2
Molecular Weight (g/mol) 225.33
MDL Number MFCD09055137
InChI Key SLGRVODTFJIPNB-UHFFFAOYSA-N
Synonym tert-butyl trans-4-vinylcyclohexyl carbamate, tert-butyl 4-ethenylcyclohexyl carbamate, tert-butyl trans-4-vinylcyclohexylcarbamate, trans-4-tert-butyl-4-vinylcyclohexyl carbamate, tert-butyl n-1r,4r-4-ethenylcyclohexyl carbamate, slgrvodtfjipnb-xypyzodxsa-n, tert-butyl 4-vinylcyclohexylcarbamate, tert-butyl 4-vinyl-cyclohexyl carbamate, trans-1-boc-amino-4-ethenylcyclohexane, trans-tert-butyl 4-vinylcyclohexylcarbamate
PubChem CID 46172842
IUPAC Name tert-butyl N-(4-ethenylcyclohexyl)carbamate
SMILES CC(C)(C)OC(=O)NC1CCC(CC1)C=C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62078MD
View Documents Promotion Details
Thermo Scientific Chemicals
H62078MD
250 mg
Each for $305.10
 
AAH6207803
View Documents Promotion Details
Thermo Scientific Chemicals
H6207803
1 g
Each for $1,102.81
 
Description

Description

trans-1-(Boc-amino)-4-vinylcyclohexane used as a intermediate in pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-1-(Boc-amino)-4-vinylcyclohexane used as a intermediate in pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
Specifications

Chemical Identifiers

1198355-16-6
225.33
SLGRVODTFJIPNB-UHFFFAOYSA-N
46172842
CC(C)(C)OC(=O)NC1CCC(CC1)C=C
C13H23NO2
MFCD09055137
tert-butyl trans-4-vinylcyclohexyl carbamate, tert-butyl 4-ethenylcyclohexyl carbamate, tert-butyl trans-4-vinylcyclohexylcarbamate, trans-4-tert-butyl-4-vinylcyclohexyl carbamate, tert-butyl n-1r,4r-4-ethenylcyclohexyl carbamate, slgrvodtfjipnb-xypyzodxsa-n, tert-butyl 4-vinylcyclohexylcarbamate, tert-butyl 4-vinyl-cyclohexyl carbamate, trans-1-boc-amino-4-ethenylcyclohexane, trans-tert-butyl 4-vinylcyclohexylcarbamate
tert-butyl N-(4-ethenylcyclohexyl)carbamate

Specifications

1198355-16-6
MFCD09055137
Slightly soluble in water.
CC(C)(C)OC(=O)NC1CCC(CC1)C=C
225.33
225.32
250 mg
C13H23NO2
tert-butyl trans-4-vinylcyclohexyl carbamate, tert-butyl 4-ethenylcyclohexyl carbamate, tert-butyl trans-4-vinylcyclohexylcarbamate, trans-4-tert-butyl-4-vinylcyclohexyl carbamate, tert-butyl n-1r,4r-4-ethenylcyclohexyl carbamate, slgrvodtfjipnb-xypyzodxsa-n, tert-butyl 4-vinylcyclohexylcarbamate, tert-butyl 4-vinyl-cyclohexyl carbamate, trans-1-boc-amino-4-ethenylcyclohexane, trans-tert-butyl 4-vinylcyclohexylcarbamate
SLGRVODTFJIPNB-UHFFFAOYSA-N
tert-butyl N-(4-ethenylcyclohexyl)carbamate
46172842
97%
trans-1-(Boc-amino)-4-vinylcyclohexane
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only