Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-1-(Boc-amino)-4-(hydroxymethyl)cyclohexane, 97%

Chemical Identifiers

Chemical Name or Material trans-1-(Boc-amino)-4-(hydroxymethyl)cyclohexane
CAS 239074-29-4
Formula Weight 229.31
MDL Number MFCD03844605
Synonym tert-Butyl trans-(4-hydroxymethyl)cyclohexylcarbamate; trans-4-(Boc-amino)cyclohexanemethanol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6248703

 
alfa aesar™
H6248703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6248706

 
alfa aesar™
H6248706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6248714

 
alfa aesar™
H6248714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 229.31
Chemical Name or Material trans-1-(Boc-amino)-4-(hydroxymethyl)cyclohexane
Physical Form Solid
Assay 97%
MDL Number MFCD03844605
CAS 239074-29-4
Synonym tert-Butyl trans-(4-hydroxymethyl)cyclohexylcarbamate; trans-4-(Boc-amino)cyclohexanemethanol
Quantity 1g

Applications
trans-1-(Boc-amino)-4-(hydroxymethyl)cyclohexane is used to prepare C-2 hydroxyethyl imidazopyrrolo pyridines as JAK1 inhibitors. It is also used to prepare Mer kinase inhibitors for treatment of pediatric acute lymphoblastic leukemia.

Solubility
Slightly Soluble (4.4 g/L) (25°C).

Notes
Store in cool, dry place in tightly closed container. Keep container tightly closed. Store away from oxidizing agent.