missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-1-(Boc-amino)-4-cyanocyclohexane, 97%

$144.34 - $431.80

Chemical Identifiers

CAS 873537-32-7
Molecular Formula C12H20N2O2
Molecular Weight (g/mol) 224.304
MDL Number MFCD11707221
InChI Key QHZJRHRMFIDRFL-UHFFFAOYSA-N
Synonym tert-butyl 4-cyanocyclohexyl carbamate, tert-butyl cis-4-cyanocyclohexylcarbamate, 4-cyano-cyclohexyl-carbamic acid tert-butyl ester, tert-butyl trans-4-cyanocyclohexylcarbamate, tert-butyl n-4-cyanocyclohexyl carbamate, tert-butyl 1r,4r-4-cyanocyclohexyl carbamate, qhzjrhrmfidrfl-mgcohnpysa-n, trans-1-boc-amino-4-cyanocyclohexane, tert-butyl trans-4-cyanocyclohexylcarbamate, trans tert-butyl 4-cyanocyclohexyl carbamate
PubChem CID 46172880
IUPAC Name tert-butyl N-(4-cyanocyclohexyl)carbamate
SMILES CC(C)(C)OC(=O)NC1CCC(CC1)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62060MD View Documents Alfa Aesar™
H62060MD
250mg Each for $144.34
 
AAH6206003 View Documents Alfa Aesar™
H6206003
1g Each for $431.80
 
Specifications

Chemical Identifiers

873537-32-7
224.304
QHZJRHRMFIDRFL-UHFFFAOYSA-N
46172880
CC(C)(C)OC(=O)NC1CCC(CC1)C#N
C12H20N2O2
MFCD11707221
tert-butyl 4-cyanocyclohexyl carbamate, tert-butyl cis-4-cyanocyclohexylcarbamate, 4-cyano-cyclohexyl-carbamic acid tert-butyl ester, tert-butyl trans-4-cyanocyclohexylcarbamate, tert-butyl n-4-cyanocyclohexyl carbamate, tert-butyl 1r,4r-4-cyanocyclohexyl carbamate, qhzjrhrmfidrfl-mgcohnpysa-n, trans-1-boc-amino-4-cyanocyclohexane, tert-butyl trans-4-cyanocyclohexylcarbamate, trans tert-butyl 4-cyanocyclohexyl carbamate
tert-butyl N-(4-cyanocyclohexyl)carbamate

Specifications

250mg
C12H20N2O2
UN3439
QHZJRHRMFIDRFL-UHFFFAOYSA-N
tert-butyl N-(4-cyanocyclohexyl)carbamate
46172880
97%
873537-32-7
MFCD11707221
tert-butyl 4-cyanocyclohexyl carbamate, tert-butyl cis-4-cyanocyclohexylcarbamate, 4-cyano-cyclohexyl-carbamic acid tert-butyl ester, tert-butyl trans-4-cyanocyclohexylcarbamate, tert-butyl n-4-cyanocyclohexyl carbamate, tert-butyl 1r,4r-4-cyanocyclohexyl carbamate, qhzjrhrmfidrfl-mgcohnpysa-n, trans-1-boc-amino-4-cyanocyclohexane, tert-butyl trans-4-cyanocyclohexylcarbamate, trans tert-butyl 4-cyanocyclohexyl carbamate
CC(C)(C)OC(=O)NC1CCC(CC1)C#N
224.304
224.29
trans-1-(Boc-amino)-4-cyanocyclohexane
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.