Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-1-(Boc-amino)-4-(4-methyl-1-piperazinyl)cyclohexane, 97%

Chemical Identifiers

Chemical Name or Material trans-1-(Boc-amino)-4-(4-methyl-1-piperazinyl)cyclohexane
CAS 1119283-74-7
Formula Weight 297.43
Synonym tert-Butyl trans-4-(4-methyl-1-piperazinyl)cyclohexylcarbamate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH62731MD

 
alfa aesar™
H62731MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6273103

 
alfa aesar™
H6273103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 297.43
Chemical Name or Material trans-1-(Boc-amino)-4-(4-methyl-1-piperazinyl)cyclohexane
Physical Form Solid
Assay 97%
CAS 1119283-74-7
Synonym tert-Butyl trans-4-(4-methyl-1-piperazinyl)cyclohexylcarbamate
Quantity 250mg