Please login to your online account to display your discounted pricing

trans-1,4-Dibromo-2-butene, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC406460250

 View more versions of this product

Catalog No. AC406460250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Crystals or Crystalline Powder
Formula Weight 213.9
Quantity 25g
Color White to beige
Melting Point 48° to 53°C
Chemical Name or Material trans-1, 4-Dibromo-2-butene
Health Hazard 1 GHS Signal Word: Danger
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Recommended Storage Refrigerator +4°C
Name Note 97%
Assay Percent Range 97%
Molecular Formula C4H6Br2
Linear Formula BrCH2CHCHCH2Br
MDL Number MFCD00000249
CAS 821-06-7
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Toxic if inhaled.

Specifications

Chemical Name or Material trans-1, 4-Dibromo-2-butene
Name Note 97%
CAS 821-06-7
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C4H6Br2
Linear Formula BrCH2CHCHCH2Br
Formula Weight 213.9
MDL Number MFCD00000249
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Toxic if inhaled.

Refrigerator +4°C

SDS & Safety Documents