missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-1,2-Dibromocyclohexane, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC112771000

 View more versions of this product

Catalog No. AC112771000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

trans-1, 2-Dibromocyclohexane
>110°C
Glass bottle
1.78
Yellow
99%
99%
C6H10Br2
05, 24
CZNHKZKWKJNOTE-PHDIDXHHSA-N
(1R,2R)-1,2-dibromocyclohexane
98506
Liquid
1.7800g/mL
Authentic
1.5510 to 1.553
145°C (100.0mmHg)
100mL
7429-37-0
C6H10Br2
MFCD00003820
+/--trans-1,2-dibromocyclohexane, 1beta,2alpha-dibromocyclohexane, 1r,2r-1,2-bis bromanyl cyclohexane, 1r,2r-1,2-dibromocyclohexane, cyclohexane,1,2-dibromo-,trans, cyclohexane,2-dibromo-, trans, trans-1,2-dibromcyclohexan, trans-1,2-dibromocyclohexane
C1CCC(C(C1)Br)Br
241.95
241.95
99%

Chemical Identifiers

7429-37-0
241.95
CZNHKZKWKJNOTE-PHDIDXHHSA-N
98506
C1CCC(C(C1)Br)Br
C6H10Br2
MFCD00003820
+/--trans-1,2-dibromocyclohexane, 1beta,2alpha-dibromocyclohexane, 1r,2r-1,2-bis bromanyl cyclohexane, 1r,2r-1,2-dibromocyclohexane, cyclohexane,1,2-dibromo-,trans, cyclohexane,2-dibromo-, trans, trans-1,2-dibromcyclohexan, trans-1,2-dibromocyclohexane
(1R,2R)-1,2-dibromocyclohexane
Safety and Handling

Safety and Handling

EINECSNumber : 231-070-2

SDS Documents