missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-1,2-Dibromocyclohexane, 99%

Manufacturer:  Alfa Aesar™ H5345614

 View more versions of this product

Catalog No. AAH5345614



Description

Description

Applications
trans-1,2-Dibromocyclohexane is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

trans-1,2-Dibromocyclohexane
>113°C (235°F)
145°C (100mmHg)
7429-37-0
MFCD00003820
CZNHKZKWKJNOTE-PHDIDXHHSA-N
(1R,2R)-1,2-dibromocyclohexane
98506
99%
1.784
1.5515
25g
C6H10Br2
+/--trans-1,2-dibromocyclohexane
C1CCC(C(C1)Br)Br
241.954
241.95

Chemical Identifiers

7429-37-0
241.954
CZNHKZKWKJNOTE-PHDIDXHHSA-N
98506
C1CCC(C(C1)Br)Br
C6H10Br2
MFCD00003820
+/--trans-1,2-dibromocyclohexane
(1R,2R)-1,2-dibromocyclohexane
Safety and Handling

Safety and Handling

EINECSNumber : 231-070-2

TSCA : No

Recommended Storage : Ambient temperatures

Documents