missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-1,2-Dibromocyclohexane, 99%

$30.05 - $321.45

Chemical Identifiers

CAS 7429-37-0
Molecular Formula C6H10Br2
Molecular Weight (g/mol) 241.954
MDL Number MFCD00003820
InChI Key CZNHKZKWKJNOTE-PHDIDXHHSA-N
Synonym +/--trans-1,2-dibromocyclohexane
PubChem CID 98506
IUPAC Name (1R,2R)-1,2-dibromocyclohexane
SMILES C1CCC(C(C1)Br)Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5345606 View Documents Alfa Aesar™
H5345606
5g Each for $30.05
 
AAH5345614 View Documents Alfa Aesar™
H5345614
25g Each for $104.15
 
AAH5345622 View Documents Alfa Aesar™
H5345622
100g Each for $321.45
 
Description

Description

Applications
trans-1,2-Dibromocyclohexane is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

7429-37-0
241.954
CZNHKZKWKJNOTE-PHDIDXHHSA-N
98506
C1CCC(C(C1)Br)Br
C6H10Br2
MFCD00003820
+/--trans-1,2-dibromocyclohexane
(1R,2R)-1,2-dibromocyclohexane

Specifications

1.784
1.5515
5g
C6H10Br2
+/--trans-1,2-dibromocyclohexane
C1CCC(C(C1)Br)Br
241.954
241.95
trans-1,2-Dibromocyclohexane
>113°C (235°F)
145°C (100mmHg)
7429-37-0
MFCD00003820
CZNHKZKWKJNOTE-PHDIDXHHSA-N
(1R,2R)-1,2-dibromocyclohexane
98506
99%
Safety and Handling

Safety and Handling

EINECSNumber : 231-070-2

TSCA : No

Recommended Storage : Ambient temperatures