missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
trans-1,2-Dibenzoylethylene, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC112580010
Specifications
trans-1, 2-Dibenzoylethylene | |
Glass bottle | |
108.0°C to 112.0°C | |
97% | |
96% min. (GC) | |
C6H5COCH=CHCOC6H5 | |
07, IV, 2578 | |
1,2-dibenzoylethene, 1,2-dibenzoylethylene, 1,4-diphenyl-2-butene-1,4-dione, 2-butene-1,4-dione, 1,4-diphenyl, dibenzoylethylene, ethylene, 1,2-dibenzoyl, trans-1,2-dibenzoylethylene, trans-1,4-diphenyl-2-butene-1,4-dione, trans-diphenacylidene, usaf nd-57 | |
WYCXGQSQHAXLPK-VAWYXSNFSA-N | |
(E)-1,4-diphenylbut-2-ene-1,4-dione | |
735960 | |
Powder, Crystals or Flakes |
Authentic | |
Undesignated | |
1g | |
959-28-4 | |
C16H12O2 | |
MFCD00003083 | |
01,195 | |
Solubility in water: insoluble | |
C1=CC=C(C=C1)C(=O)C=CC(=O)C2=CC=CC=C2 | |
236.27 | |
236.27 | |
97% |
Chemical Identifiers
959-28-4 | |
236.27 | |
WYCXGQSQHAXLPK-VAWYXSNFSA-N | |
735960 | |
C1=CC=C(C=C1)C(=O)C=CC(=O)C2=CC=CC=C2 |
C16H12O2 | |
MFCD00003083 | |
1,2-dibenzoylethene, 1,2-dibenzoylethylene, 1,4-diphenyl-2-butene-1,4-dione, 2-butene-1,4-dione, 1,4-diphenyl, dibenzoylethylene, ethylene, 1,2-dibenzoyl, trans-1,2-dibenzoylethylene, trans-1,4-diphenyl-2-butene-1,4-dione, trans-diphenacylidene, usaf nd-57 | |
(E)-1,4-diphenylbut-2-ene-1,4-dione |
Safety and Handling
EINECSNumber : 213-498-1
TSCA : TSCA