Please login to your online account to display your discounted pricing

trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic Acid, Monohydrate, Macron Fine Chemicals™


macron

Quantity 25g

Manufacturer: avantor performance  235755

 View more versions of this product

Catalog No. MK235725

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 25g
Chemical Name or Material trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic Acid, Monohydrate
Assay 98% min.
Molecular Formula C14H22N2O8·H2O
Formula Weight 364.36
CAS 13291-61-7
Synonym DCYTA

Specifications

Chemical Name or Material trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic Acid, Monohydrate
CAS 13291-61-7
Molecular Formula C14H22N2O8·H2O
Formula Weight 364.36
Synonym DCYTA