Learn More
trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic Acid Monohydrate, 98%, Thermo Scientific Chemicals
trans-1,2-Diaminocyclohexane-N,N,N,N-tetraacetic acid monohydrate, 98%, C14H24N2O9, CAS Number-125572-95-4
Supplier: Thermo Scientific Chemicals 155005000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagentSpecifications
trans-1,2-Diaminocyclohexane-N,N,N′,N′-tetraacetic acid monohydrate | |
>210.0°C | |
Authentic | |
Plastic bottle | |
C6H10[N(CH2CO2H)2]2·H2O | |
500 g | |
2,2',2,2'-trans-cyclohexane-1,2-diylbis azanetriyl tetraacetic acid hydrate, dcyta, trans-1,2-diaminocyclohexane-n,n,n',n'-tetraacetic acid monohydrate, 1,2-cyclohexanedinitrilotetraacetic acid, 2,2',2,2'-1r,2r-rel-cyclohexane-1,2-diylbis azanetriyl tetraacetic acid hydrate, trans-1,2-cyclohexanediaminetetraacetic acid monohydrate, cdta monohydrate, ctda monohydrate, chel-cd, chel r-cd | |
FCKYPQBAHLOOJQ-NXEZZACHSA-L | |
2-[[(1R,2R)-2-[bis(carboxymethyl)amino]cyclohexyl]-(carboxymethyl)amino]acetic acid;hydrate | |
2723844 | |
98% |
125572-95-4 | |
White | |
97.5% min. (Complexometry) | |
C14H20N2O8 | |
MFCD00149243,MFCD00066429,MFCD00003845 | |
13,III,1 | |
Solubility in water: insoluble. Other solubilities: insoluble in most common organic solvents, soluble in alkali solutions | |
[O-]C(=O)C[NH+](CC([O-])=O)[C@@H]1CCCC[C@H]1[NH+](CC([O-])=O)CC([O-])=O | |
344.32 | |
364.35 | |
Fine Crystalline Powder |
Chemical Identifiers
125572-95-4 | |
344.32 | |
FCKYPQBAHLOOJQ-NXEZZACHSA-L | |
2723844 |
C14H20N2O8 | |
MFCD00149243,MFCD00066429,MFCD00003845 | |
2,2',2,2'-trans-cyclohexane-1,2-diylbis azanetriyl tetraacetic acid hydrate, dcyta, trans-1,2-diaminocyclohexane-n,n,n',n'-tetraacetic acid monohydrate, 1,2-cyclohexanedinitrilotetraacetic acid, 2,2',2,2'-1r,2r-rel-cyclohexane-1,2-diylbis azanetriyl tetraacetic acid hydrate, trans-1,2-cyclohexanediaminetetraacetic acid monohydrate, cdta monohydrate, ctda monohydrate, chel-cd, chel r-cd | |
[O-]C(=O)C[NH+](CC([O-])=O)[C@@H]1CCCC[C@H]1[NH+](CC([O-])=O)CC([O-])=O |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre
GHS Signal Word: Warning
RUO – Research Use Only