missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(±)-trans-1,2-Diaminocyclohexane, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC248672500

 View more versions of this product

Catalog No. AC248672500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

(±)-trans-1,2-Diaminocyclohexane
68°C
Glass bottle
0.951
Colorless to Yellow
250mL
1121-22-8
100.0
C6H14N2
MFCD00063747
Solubility in water: soluble.
C1CCC(C(C1)N)N
114.19
114.19
98%
0.9510g/mL
Authentic
1.4885 to 1.4905
79.0°C to 81.0°C (15.0mmHg)
14.0°C to 15.0°C
98%
97.5
97.5% min. (GC)
C6H10(NH2)2
1r,2r---1,2-cyclohexanediamine, 1r,2r---1,2-diaminocyclohexane, 1r,2r-1,2-cyclohexanediamine, 1r,2r-1,2-diaminocyclohexane, 1r,2r-cyclohexane-1,2-diamine, 1r,2r-diaminocyclohexane, r-dach, trans-1,2-cyclohexanediamine, trans-1,2-diaminocyclohexane, trans-cyclohexane-1,2-diamine
SSJXIUAHEKJCMH-PHDIDXHHSA-N
(1R,2R)-cyclohexane-1,2-diamine
43806
Liquid

Chemical Identifiers

1121-22-8
114.19
SSJXIUAHEKJCMH-PHDIDXHHSA-N
43806
C1CCC(C(C1)N)N
C6H14N2
MFCD00063747
1r,2r---1,2-cyclohexanediamine, 1r,2r---1,2-diaminocyclohexane, 1r,2r-1,2-cyclohexanediamine, 1r,2r-1,2-diaminocyclohexane, 1r,2r-cyclohexane-1,2-diamine, 1r,2r-diaminocyclohexane, r-dach, trans-1,2-cyclohexanediamine, trans-1,2-diaminocyclohexane, trans-cyclohexane-1,2-diamine
(1R,2R)-cyclohexane-1,2-diamine
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents