missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

trans-1,2-Bis(tributylstannyl)ethylene, TCI America™

$131.39 - $397.23

Chemical Identifiers

CAS 14275-61-7
Molecular Formula C26H56Sn2
Molecular Weight (g/mol) 606.154
MDL Number MFCD01631299
InChI Key VNKOWRBFAJTPLS-UHFFFAOYSA-N
Synonym trans-1,2-bis tributylstannyl ethene, e-1,2-bis tributylstannyl ethene, trans-1,2-bis tri-n-butylstannyl ethylene, tributyl e-2-tributylstannyl ethenyl stannane, stannane, 1e-1,2-ethenediylbis tributyl, stannane, 1,2-ethenediylbis tributyl, trans-1,2-bis tri-n-butylstannyl ethylene, e-1,2-bis-tri-n-butylstannyl-ethylene, trans-1,2-bis tributylstannyl ethene, tributyl 2-tributylstannyl ethenyl stannane
PubChem CID 5378370
IUPAC Name tributyl-[(E)-2-tributylstannylethenyl]stannane
SMILES CCCC[Sn](CCCC)(CCCC)C=C[Sn](CCCC)(CCCC)CCCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B19755G View Documents TCI America
B19755G
5g Each for $397.23
 
B19751G View Documents TCI America
B19751G
1g Each for $131.39
 
Specifications

Chemical Identifiers

14275-61-7
606.154
VNKOWRBFAJTPLS-UHFFFAOYSA-N
5378370
CCCC[Sn](CCCC)(CCCC)C=C[Sn](CCCC)(CCCC)CCCC
C26H56Sn2
MFCD01631299
trans-1,2-bis tributylstannyl ethene, e-1,2-bis tributylstannyl ethene, trans-1,2-bis tri-n-butylstannyl ethylene, tributyl e-2-tributylstannyl ethenyl stannane, stannane, 1e-1,2-ethenediylbis tributyl, stannane, 1,2-ethenediylbis tributyl, trans-1,2-bis tri-n-butylstannyl ethylene, e-1,2-bis-tri-n-butylstannyl-ethylene, trans-1,2-bis tributylstannyl ethene, tributyl 2-tributylstannyl ethenyl stannane
tributyl-[(E)-2-tributylstannylethenyl]stannane

Specifications

Yellow
14275-61-7
MFCD01631299
trans-1,2-bis tributylstannyl ethene, e-1,2-bis tributylstannyl ethene, trans-1,2-bis tri-n-butylstannyl ethylene, tributyl e-2-tributylstannyl ethenyl stannane, stannane, 1e-1,2-ethenediylbis tributyl, stannane, 1,2-ethenediylbis tributyl, trans-1,2-bis tri-n-butylstannyl ethylene, e-1,2-bis-tri-n-butylstannyl-ethylene, trans-1,2-bis tributylstannyl ethene, tributyl 2-tributylstannyl ethenyl stannane
CCCC[Sn](CCCC)(CCCC)C=C[Sn](CCCC)(CCCC)CCCC
606.154
606.15
≥98.0% (W)
5g
C26H56Sn2
2788
VNKOWRBFAJTPLS-UHFFFAOYSA-N
tributyl-[(E)-2-tributylstannylethenyl]stannane
5378370
Clear Liquid at 20°C
trans-1,2-Bis(tributylstannyl)ethylene
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator