missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-1,2-Bis(tri-n-butylstannyl)ethylene, 96%

Chemical Identifiers

CAS 14275-61-7
Molecular Formula C26H56Sn2
Formula Weight 606.15
MDL Number MFCD01631299
UN Number UN2788
Beilstein 3613803
Chemical Name or Material trans-1,2-Bis(tri-n-butylstannyl)ethylene
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL2013003 SDS Alfa Aesar™
L2013003
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAL2013006 SDS Alfa Aesar™
L2013006
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Applications
trans-1,2-Bis(tri-n-butylstannyl)ethylene is used as an intermediate in organic synthesis. It is also employed in the synthesis of gamma, delta-alkynones.

Solubility
Miscible with terahydrofuran and dimethyl formamide. Slightly miscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Description & Specifications

Chemical Identifiers

14275-61-7
606.15
UN2788
trans-1,2-Bis(tri-n-butylstannyl)ethylene
C26H56Sn2
MFCD01631299
3613803

Specifications

14275-61-7
606.15
UN2788
185°C (0.01mmHg)
trans-1,2-Bis(tri-n-butylstannyl)ethylene
C26H56Sn2
MFCD01631299
3613803
1g
96%

Safety and Handling

GHS H Statement
H300-H372-H312-H315-H319
Fatal if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P260-P280-P301+P310-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

Recommended Storage : 2° to 8°C