Learn More
trans-1,2-Bis(tri-n-butylstannyl)ethylene, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L2013006
Description
trans-1,2-Bis(tri-n-butylstannyl)ethylene is used as an intermediate in organic synthesis. It is also employed in the synthesis of gamma, delta-alkynones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationstrans-1,2-Bis(tri-n-butylstannyl)ethylene is used as an intermediate in organic synthesis. It is also employed in the synthesis of gamma, delta-alkynones.
Solubility
Miscible with terahydrofuran and dimethyl formamide. Slightly miscible with water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
trans-1,2-Bis(tri-n-butylstannyl)ethylene | |
1.14 | |
C26H56Sn2 | |
MFCD01631299 | |
UN2788 | |
Air and moisture sensitive | |
Miscible with terahydrofuran and dimethyl formamide. Slightly miscible with water. | |
CCCC[Sn](CCCC)(CCCC)C=C[Sn](CCCC)(CCCC)CCCC | |
606.154 | |
606.15 |
14275-61-7 | |
185°C (0.01 mmHg) | |
1.502 | |
5 g | |
3613803 | |
trans-1,2-bis tributylstannyl ethene, e-1,2-bis tributylstannyl ethene, trans-1,2-bis tri-n-butylstannyl ethylene, tributyl e-2-tributylstannyl ethenyl stannane, stannane, 1e-1,2-ethenediylbis tributyl, stannane, 1,2-ethenediylbis tributyl, e-1,2-bis-tri-n-butylstannyl-ethylene, tributyl 2-tributylstannyl ethenyl stannane | |
VNKOWRBFAJTPLS-UHFFFAOYSA-N | |
tributyl-[(E)-2-tributylstannylethenyl]stannane | |
5378370 | |
96% |
Chemical Identifiers
14275-61-7 | |
606.154 | |
VNKOWRBFAJTPLS-UHFFFAOYSA-N | |
5378370 | |
CCCC[Sn](CCCC)(CCCC)C=C[Sn](CCCC)(CCCC)CCCC |
C26H56Sn2 | |
MFCD01631299 | |
trans-1,2-bis tributylstannyl ethene, e-1,2-bis tributylstannyl ethene, trans-1,2-bis tri-n-butylstannyl ethylene, tributyl e-2-tributylstannyl ethenyl stannane, stannane, 1e-1,2-ethenediylbis tributyl, stannane, 1,2-ethenediylbis tributyl, e-1,2-bis-tri-n-butylstannyl-ethylene, tributyl 2-tributylstannyl ethenyl stannane | |
tributyl-[(E)-2-tributylstannylethenyl]stannane |
Safety and Handling
GHS H Statement
H300-H372-H312-H315-H319
Fatal if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
H301-H312-H315-H319-H372
DOTInformation : Hazard Class: 6.1; Packaging Group: III
TSCA : No
Recommended Storage : Keep cold; Store under Argon; Store under Nitrogen
RUO – Research Use Only