Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-1,2-Bis(diphenylphosphino)ethylene, 98%

$244.74 - $244.74

Chemical Identifiers

Chemical Name or Material trans-1,2-Bis(diphenylphosphino)ethylene
CAS 983-81-3
Molecular Formula C26H22P2
Formula Weight 396.41
MDL Number MFCD00063219
Beilstein 757793
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0850806

  SDS
alfa aesar™
L0850806
5g Each for $244.74
Description & Specifications

Specifications

Formula Weight 396.41
Chemical Name or Material trans-1,2-Bis(diphenylphosphino)ethylene
Molecular Formula C26H22P2
CAS 983-81-3
Assay 98%
MDL Number MFCD00063219
Beilstein 757793
Melting Point 126° to 130°C
Health Hazard 1 Warning
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.