Please login to your online account to display your discounted pricing

trans-1,2-Bis(diphenylphosphino)ethylene, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material trans-1, 2-Bis(diphenylphosphino)ethylene
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C26 H22 P2
Linear Formula (C6H5)2PCH=CHP(C6H5)2
Formula Weight 396.41
MDL Number MFCD00063219
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC310040010

  SDS
thermo fisher scientific
310040010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Fine Crystalline Powder
Formula Weight 396.41
Quantity 1g
Color White
Melting Point 123° to 128°C
Chemical Name or Material trans-1, 2-Bis(diphenylphosphino)ethylene
Packaging Glass bottle
Assay Percent Range 97%
CAS 983-81-3
Infrared Spectrum Authentic
Molecular Formula C26 H22 P2
MDL Number MFCD00063219
Linear Formula (C6H5)2PCH=CHP(C6H5)2
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.