Please login to your online account to display your discounted pricing

trans-1,2-Bis(diphenylphosphino)ethylene, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  310040010

 View more versions of this product

Catalog No. AC310040010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Fine Crystalline Powder
Formula Weight 396.41
Quantity 1g
Color White
Melting Point 123° to 128°C
Packaging Glass bottle
Assay Percent Range 97%
CAS 983-81-3
Chemical Name or Material trans-1, 2-Bis(diphenylphosphino)ethylene
Molecular Formula C26 H22 P2
MDL Number MFCD00063219
Infrared Spectrum Authentic
Linear Formula (C6H5)2PCH=CHP(C6H5)2
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material trans-1, 2-Bis(diphenylphosphino)ethylene
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C26 H22 P2
Linear Formula (C6H5)2PCH=CHP(C6H5)2
Formula Weight 396.41
MDL Number MFCD00063219
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents