Please login to your online account to display your discounted pricing

Alfa Aesar™ trans-1,2-Bis(4-fluorobenzoyl)ethylene, 98%

$51.42 - $137.14

Chemical Identifiers

Chemical Name or Material trans-1,2-Bis(4-fluorobenzoyl)ethylene
CAS 25650-13-9
Molecular Formula C16H10F2O2
Formula Weight 272.25
MDL Number MFCD00051604
Beilstein 1979161
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0619303

  SDS
alfa aesar™
L0619303
1g Each for $137.14

AAL06193MD

  SDS
alfa aesar™
L06193MD
250mg Each for $51.42
Description & Specifications

Specifications

Formula Weight 272.25
Chemical Name or Material trans-1,2-Bis(4-fluorobenzoyl)ethylene
Assay 98%
MDL Number MFCD00051604
CAS 25650-13-9
Beilstein 1979161
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C16H10F2O2
Melting Point 161° to 163°C
Health Hazard 1 Warning
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.