Please login to your online account to display your discounted pricing

Tranilast, Tocris


Antiallergic; inhibits inflammatory mediator release from mast cells

Quantity 10mg

Manufacturer: r & d systems  RDS109810

 View more versions of this product

Catalog No. 109810

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 327.34
Quantity 10mg
Chemical Name or Material Tranilast
CAS 53902-12-8
Synonym 2-[[3-(3, 4-Dimethoxyphenyl)-1-oxo-2-propenyl]amino]benzoic acid
Recommended Storage Store at Room Temperature
Assay Percent Range >99%
Molecular Formula C18H17NO5

Specifications

Chemical Name or Material Tranilast
Assay Percent Range >99%
Molecular Formula C18H17NO5
Formula Weight 327.34
Synonym 2-[[3-(3, 4-Dimethoxyphenyl)-1-oxo-2-propenyl]amino]benzoic acid
Accessories

Store at Room Temperature