Please login to your online account to display your discounted pricing

TRAM 34, Tocris

Highly selective KCa3.1 channel blocker

$164.61 - $682.67

Chemical Identifiers

Chemical Name or Material TRAM 34
Assay Percent Range >99%
Molecular Formula C22H17ClN2
Formula Weight 344.84
Synonym 1-[(2-Chlorophenyl)diphenylmethyl]-1H-pyrazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

294610

 
r & d systems
294610
10mg Each for $164.61

294650

 
r & d systems
294650
50mg Each for $682.67
Description & Specifications

Specifications

Chemical Name or Material TRAM 34
Quantity 10mg
CAS 289905-88-0
Recommended Storage Store at +4°C
Assay Percent Range >99%
Molecular Formula C22H17ClN2
Formula Weight 344.84
Synonym 1-[(2-Chlorophenyl)diphenylmethyl]-1H-pyrazole
Safety and Handling

Store at +4°C