missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tralkoxydim, TCI America™

Manufacturer:  TCI America T29161G

 View more versions of this product

Catalog No. T29161G



Specifications

Specifications

Tralkoxydim
107°C
87820-88-0
MFCD00145415
SOTLWPHEAQOHHC-UHFFFAOYSA-N
2-[1-(ethoxyamino)propylidene]-5-(2,4,6-trimethylphenyl)cyclohexane-1,3-dione
91746
Crystal-Powder at 20°C
Undesignated
1g
C20H27NO3
3077
CCC(=C1C(=O)CC(CC1=O)C2=C(C=C(C=C2C)C)C)NOCC
329.44
329.44
≥95.0% (HPLC,N)

Chemical Identifiers

87820-88-0
329.44
SOTLWPHEAQOHHC-UHFFFAOYSA-N
2-[1-(ethoxyamino)propylidene]-5-(2,4,6-trimethylphenyl)cyclohexane-1,3-dione
C20H27NO3
MFCD00145415
91746
CCC(=C1C(=O)CC(CC1=O)C2=C(C=C(C=C2C)C)C)NOCC
Safety and Handling

Safety and Handling

RTECSNumber : GW7191600

TSCA : No

Documents