Please login to your online account to display your discounted pricing

TRAF-1 Rat anti-Human, Clone: 1F3, eBioscience™

Rat Monoclonal Antibody

$175.45 - $442.25

Specifications

Antigen TRAF-1
Clone 1F3
Host Species Rat
Gene Alias TNF receptor-associated factor 1, EBI6, Epstein-Barr virus-induced protein 6
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122871

 
life technologies
LS14979180
25μg Each for $175.45

501122872

 
life technologies
LS14979182
100μg Each for $442.25
Description & Specifications

Specifications

Antigen TRAF-1
Clone 1F3
Host Species Rat
Gene Alias TNF receptor-associated factor 1, EBI6, Epstein-Barr virus-induced protein 6
Species Reactivity Human
Applications Immunocytochemistry
Applications Immunohistochemistry
Applications Microscopy
Applications Western Blot
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The monoclonal antibody 1F3 recognizes human TRAF-1. TRAF-1 is a multifunctional adapter molecule and member of the TNF receptor associated factor protein family. TRAF proteins associate with various receptors of the TNF receptor superfamily to mediate signal transduction. TRAF-1 plays a role in regulating cell survival and apoptosis, regulates the signaling of NF kappa B and JNK, and forms a heterodimeric complex with TRAF-2. TRAF-1 also interacts with cell surface receptors such as CD30, CD40, and CD357 (AITR), and cytoplasmic proteins like NIK, RIP2, A20, and TRIP. Expression of TRAF-1 can be induced by Epstein-Barr Virus (EBV) and TNF alpha.