missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ Yttrium Standard for AAS, 1,000 mg/L Y in Nitric Acid, MilliporeSigma™ Supelco™
Specifications
Specifications
| Product Line | TraceCERT |
| CAS | 7697-37-2 |
| Type | Certified Reference Material |
| Quantity | 250 mL |
| Grade | TraceCERT™ Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 1,000 mg/L Y in nitric acid |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC