missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ Ethylene Oxide Solution, Certified Reference Material, 50 mg/mL in Dichloromethane, MilliporeSigma™ Supelco™
Specifications
Specifications
| Packaging | Ampule |
| Product Line | TraceCERT |
| Concentration | 50 mg/mL in dichloromethane |
| Type | Organic Pollutant Standard |
| Quantity | 1 mL |
| Grade | TraceCERT™ Analytical Standard |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC