Please login to your online account to display your discounted pricing

Fisher Scientific™ Traceable™ Excursion-Trac™ Datalogging Thermometers: PROMO OFFER

PROMOTIONAL OFFER: Buy a Fisher Scientific Traceable Excursion-Trac Datalogging Thermometer and get a Fisher Scientific Traceable Radio Atomic Clock (15-076-6) at no additional cost

$246.07 - $264.52

Specifications

Product Type Datalogging Thermometer
View More Specs
Includes:  Stand, Velcro™, magnetic strips, wall mount, batteries, and an ISO 17025 Traceable™ Certificate
Products
Catalog Number Mfr. No. Description Price Quantity    

15-081-123PMO

 
fisher scientific™
CON6430PMO
1 Bottle Probe Each for $246.07

15-081-124PMO

 
fisher scientific™
CON6431PMO
2 Bottle Probes Each for $264.52
Description & Specifications

Specifications

Product Type Datalogging Thermometer

PROMOTIONAL OFFER: Buy this Fisher Scientific™ Traceable™ Excursion-Trac™ Datalogging Thermometer and get a Fisher Scientific™ Traceable™ Radio Atomic Clock FREE.  Offer valid through February 28, 2016.

Datalogging thermometer with user-defined time intervals.

High-accuracy thermometer has a temperature range of -50 to 70°C and resolution of 0.01 and maintains an accuracy of ±0.25°C across the entire temperature range. Ideal for monitoring temperatures in refrigerators, freezers, water baths, heating blocks, and incubators.

 

  • Stores up to ten unique alarm events
  • Meets current CDC requirements for vaccine storage and monitoring
  • Rolling memory structure, maintains most recent 525,600 temperature observations
  • Memory can be cleared on unit after downloading data to USB stick
  • Hassle-free retrieval of data; unit can remain in use while downloading and analyzing data
  • Status indicators: Low battery, memory full, USB data transfer and active alarm state
  • Traceable to NIST for accuracy
  • High-impact, chemical-resistant ABS plastic case measures 7 x 10.8 x 1.9cm (2.75 x 4.25 x 0.75 in.)
  • Weight: 5 oz.
Hi/Lo Alarms and Time/Date Stamps
  • Alarm tracking feature stores in memory each alarm event detailing when the alarm state occurred, as well as when the unit returned back to within range
  • Alarm state indicators include visual LEDs, audio alerts, and flashing LCD segments
  • Data for up to ten different alarm events can be retrieved with the most current event viewable on the display
USB Data Transferring
  • New feature allows recorded data (CSV file) to be transferred from thermometer to PC or Mac™ using a USB flash drive (not included)
  • No software is needed for computer interface
  • Each thermometer has a unique ID allowing multiple units to be used in the same location
Data-Logging Features
  • Monitor readings overnight, on weekends, or for any time period with rolling data log
  • User-defined data-logging interval of one observation/minute to one observation/24 hours
  • Memory capacity maintains a year of recorded data when using one-minute intervals and longer when using larger intervals
Bottle Probe Features
  • Bottle probes are sealed in a miniature bottle (1 x 2.5 in.) filled with nontoxic glycol
  • Solution is GRAS (generally recognized as safe) by the FDA (Food and Drug Administration)
  • Eliminates concerns about incidental contact with food or drinking water
  • Temperature-buffered bottle sensor eliminates rapid changes when refrigerator door is opened
  • Ideal for use in monitoring storage conditions for vaccines or other important samples

Replacement battery: Cat. No. 1111

Supplied with an ISO 17025 Traceable™ Certificate